Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Ictotest™ Reagent Tablets

Catalog No. am2591 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
100 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM-2591 100 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AM-2591 Supplier Siemens Healthineers Supplier No. 10337316
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Detect bilirubin in urine

  • Bottle of 100 tablets

TRUSTED_SUSTAINABILITY

Specifications

Type Ictotest™ Reagent tTblet
For Use With (Application) Detect bilirubin in Urine
Quantity 100 Tablets
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.