Learn More
Siemens Healthineers HEMA-TEK 2000™ Slide Stainer
Shop All Siemens Healthineers Products
Click to view available options
Product Type:
Hematek 2000 Slide Stainer
Description
- Pumps deliver fresh reagents in precise volume for every stain while the platen provides uniform stain from field-to-field and slide-to-slide with stain and buffer mixing accurately and uniformly
- Slides are immediately available for viewing upon completion of staining, and the rinse reagent contains the optimal alcohol concentration for proper slide drying
- Dials, located on front panel, allow for wide variation in stain intensities to suit individual needs
- Simple to maintain
- Stains 60 slides per hour
Specifications
Specifications
| Product Type | Hematek 2000 Slide Stainer |
| Slide Capacity | 60 |