Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Hematek™ 3000 System

Catalog No. 23044132 Shop All Siemens Healthineers Products
Click to view available options
Product Type:
Hematek 3000 System

Practical automation in slidestaining for laboratories of all sizes

  • Precise technology for precise slide staining
  • Provides semiautomated slide staining for laboratories that need a robust system
  • Capable of staining up to 60 slides per hour
  • Helps maximize productivity and deliver quality results through user-defined features and precision controls
  • Continuous-feed loading platform eliminates wasted time associated with batch staining methods
  • Pumps deliver a fresh, precise volume of reagent for every slide,
  • Platen provides uniform stain from field to field and slide to slide with accurate mixing and uniform stain and buffer delivery
  • Individual, consistent slide staining optimizes staining quality and enables user-adaptable intensity for standardized slide staining
  • Slides are immediately available for viewing upon completion of staining
  • Rinse reagent contains the optimal alcohol concentration for proper slide drying
  • Practical technology leads to practical advancements
  • Built on the proven technology of its predecessors, while new features further support easy and environmentally friendly features
  • Simple pump release mechanism streamlines maintenance
  • System warnings alert users of low stain-level and full waste bin to help ensure uninterrupted, quality results
  • Minimal system maintenance helps maximize uptime in your lab
  • Compact, RoHS-compliant system guarantees incorporation of environmentally friendly, sustainable substances and supports “green” initiatives

  • Processing system: Platen
  • Power requirements: 110–230 Volts
  • Dryer:Yes
  • Temperature: Room temperature
  • Cleaning agent: Methanol
  • Waste disposal:Self-contained waste drawer
  • Weight: kg (lb) 20 (44)
  • Dimensions: (H×W×D), CM (IN.) 22×54×46 (8.7×21.3×18.1)
  • Slide collection drawer included
  • Manufacturer's reagents: Hematek Modified Wright's Stain, Hematek Wright-Giemsa Stain
  • Stained slide dryer included
  • Carrier, continuous load: Spiral gear
  • Slides/hr: ∼60
  • Cellular applications: Peripheral blood and bone marrow smears

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hematek 3000 System
Certifications/Compliance Meets requirements of TUV and UL
For Use With (Application) Hematology laboratories (Peripheral blood and bone marrow smears)
Includes Stained slide dryer
Weight (English) 44 lb.
Weight (Metric) 20 kg
Height (English) 8.7 in.
Height (Metric) 22 cm
Width (English) 21.3 in.
Width (Metric) 54 cm
Voltage 110/230 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.