Learn More
DuPont™ Sierra™ Cleanroom Face Veil
Catalog No. 189992904 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Dspsble Face Veil
Catalog No.
18-999-2904 Dspsble Face Veil
1 options
This item is not returnable. View return policy
Catalog No. 18-999-2904 Supplier DuPont™ Supplier No. PP7400WH000150BH
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Cleanroom face veil combines high breathability with exceptional filtration efficiency.
  • Garments are gamma sterilized to SAL of 10-6
  • Mask is 7 in. size with bound Tyvek ties for cleanroom use
  • Pleated rayon outer facing
  • Metal nose piece
  • Adhesive at nose
  • One size fits most
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Veil
Color White
Fastening Type Snaps
Size Standard
Disposable Single-use
Model PP7400WH000150BH