Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144541 Supplier Invitrogen™ Supplier No. PA5144541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIGF Polyclonal Antibody for Western Blot, ELISA

Sigma factors are initiation factors that promote the attachment of plastid-encoded RNA polymerase (PEP) to specific initiation sites and are then released. Regulates transcription in chloroplast in a DG1-dependent manner. Involved in light-dependent chloroplast development. Required during early plant development and primary leaf formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.219 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SIGF
Gene Accession No. Q9LD95
Gene Alias SIG6; SOLDAT8
Gene Symbols SIGF
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana SIGF protein (296-547aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 818273
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.