Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGIRR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520078
Description
A suggested positive control is A549 cell lysate. PA5-20078 can be used with blocking peptide PEP-0196.
SIGIRR (single Ig IL-1R-related molecule) is a novel member of the interleukin 1 receptor (IL-1R) superfamily. Although it shows the typical conserved motifs that characterize the IL-1R and Toll superfamily, it is structurally and functionally distinct from both. SIGIRR has only one Ig domain in its extracellular portion whereas the IL-1R family contains three Ig folds. An unusually long cytoplasmic domain is reminiscent of the structure of drosophila Toll, yet the SIGIRR peptide sequence is more closely related to IL-1RI. A chimera, in which the SIGIRR intracellular domain was fused to the IL-1R extracellular domain, did not activate NFkappaB unlike similar fusion proteins of other IL-1R related molecules. Phylogenetic data that suggest that the plasma membrane protein SIGIRR is a close functional relative of MyD88 in mammals.
Specifications
SIGIRR | |
Polyclonal | |
Unconjugated | |
SIGIRR | |
AI256711; IL-1R8; MGC110992; Sigirr; single Ig and TIR domain containing; single Ig IL-1 receptor related protein; single Ig IL-1-related receptor; Single Ig IL-1R-related molecule; single Ig IL-1R-related protein; single immunoglobulin and toll-interleukin 1 receptor (TIR) domain; single immunoglobulin domain IL1R1 related; single immunoglobulin domain-containing IL1R-related protein; TIR8; Toll/interleukin-1 receptor 8; UNQ301/PRO342 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24058, 59307 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6IA17, Q9JLZ8 | |
SIGIRR | |
A synthetic peptide corresponding to 15 amino acids at the C-terminus of mouse SIGIRR. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction