Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-1/CD169 Antibody (908102), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5197MFV500
Description
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Siglec-1/CD169 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 6614 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 908102 | |
| Flow Cytometry, CyTOF-ready | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-1/CD169, Ser20-Gln1641, Accession # Q9BZZ2 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction