Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280947
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human, Mouse, Porcine, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay, CyTOF-ready.Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot reported in scientific literature, Flow Cytometry : 1:10-1:1000, ELISA : 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28794041), Immunohistochemistry-Paraffin : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, In vitro assay reported in scientific literature (PMID 18414664), Radioimmunoassay, CyTOF-ready | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Porcine reactivity reported in scientific literature (PMID: 27441395). Virus reactivity reported in scientific literature (PMID: 27487279). Mouse reactivity reported in scientific literature. Not yet tested in other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Radioimmune Assays (RIA), CyTOF | |
| HSn 7D2 | |
| Unconjugated | |
| PBS | |
| SIGLEC1 | |
| Full-length purified human Sialoadhesin (d1-4)Fc protein. [Swiss-Prot# Q9BZZ2] | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 6614 | |
| Human, Mouse, Pig, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction