Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308615MFV500
Description
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Human samples. It is validated for Flow CytometrySpecifications
Siglec-1/CD169 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
HSn 7D2 | |
Flow Cytometry | |
CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
Fc fusion protein containing N-terminal 4 domains of human Siglec-1/CD169. (Uniprot: Q9ZZ2) | |
0.1 mL | |
Adaptive Immunity, Immunology | |
6614 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction