Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600534PECY55
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human, Mouse, Porcine, Virus samples. It is validated for Flow Cytometry.Specifications
Siglec-1/CD169 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
SIGLEC1 | |
Full-length purified human Sialoadhesin (d1-4)Fc protein. [Swiss-Prot No. Q9BZZ2] | |
0.1 mL | |
Primary | |
Human, Mouse, Pig, Virus | |
IgG1 |
Flow Cytometry | |
HSn 7D2 | |
Flow Cytometry | |
CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Immunology | |
6614.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction