Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-10 Antibody (5G6), FITC, Novus Biologicals™
SDP

Catalog No. NB260457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260457 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260457 Supplier Novus Biologicals Supplier No. NBP321494F
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Siglec-10
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5G6
Conjugate FITC
Dilution ELISA, Immunohistochemistry
Formulation PBS
Gene Alias MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7
Host Species Mouse
Immunogen Siglec-10 extracellular domain fused with human IgG1 Fc fragment
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 89790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.