Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-10 Antibody (5G6), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321494JF646
Description
Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Siglec-10 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
5G6 | |
ELISA, Immunohistochemistry | |
MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7 | |
Siglec-10 extracellular domain fused with human IgG1 Fc fragment | |
0.1 mL | |
Immunology | |
89790 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction