Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$428.79 - $684.12
Specifications
| Antigen | Siglec-11 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Siglec-11 Polyclonal antibody specifically detects Siglec-11 in Human samples. It is validated for ImmunofluorescenceSpecifications
| Siglec-11 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS, pH 7.2, 40% glycerol | |
| 114132 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| sialic acid binding Ig-like lectin 11, sialic acid-binding Ig-like lectin 11, Sialic acid-binding lectin 11, siglec-11 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YSEIKIHTGQPLRGPGFGLQLEREMSGMVPK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title