Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-14 Antibody (2457E), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB4905L
Description
Siglec-14 Monoclonal antibody specifically detects Siglec-14 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Siglec-14 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
100049587 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, CyTOF | |
2457E | |
Flow Cytometry, CyTOF-ready | |
AC018755.2, sialic acid binding Ig-like lectin 14 | |
Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction