Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-14 Antibody (2457E), PE/Atto594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB4905PEATT594
Description
Siglec-14 Monoclonal antibody specifically detects Siglec-14 in Human samples. It is validated for Flow CytometrySpecifications
Siglec-14 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
100049587 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Flow Cytometry | |
2457E | |
Flow Cytometry | |
AC018755.2, sialic acid binding Ig-like lectin 14 | |
Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction