Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-16 Antibody (706045), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB68191K
Description
Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Siglec-16 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
400709 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
706045 | |
Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Siglec-P16, SIGLECP16 | |
Human Siglec-16 peptide, Accession # A6NMB1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction