Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-2/CD22 Antibody (087), DyLight 680

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290807FR
Description
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Mouse samples. It is validated for Western Blot.Specifications
Siglec-2/CD22 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
87 | |
Western Blot | |
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Siglec-2/CD22 (BAE33829.1; Met1-Arg708). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology, Phospho Specific | |
933 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction