Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-2/CD22 Antibody (219934), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1968MFV500
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| Siglec-2/CD22 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology, Phospho Specific | |
| 933 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 219934 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-2/CD22, Asp20-Arg687, Accession # CAA42006 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction