Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-2/CD22 Antibody (TPD44/D11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Siglec-2/CD22 |
---|---|
Clone | TPD44/D11 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Siglec-2/CD22 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology | |
PBS | |
933 | |
IgG2a | |
Protein A purified |
TPD44/D11 | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
B cell lymphoma cells | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title