Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-2/CD22 Antibody (OTI1F12), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274186IR
Description
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Siglec-2/CD22 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD22 | |
Full length recombinant protein of human Siglec-2/CD22 (NP_001762) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1F12 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
Mouse | |
Affinity Purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology, Phospho Specific | |
933.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction