Learn More
Siglec-3/CD33 Antibody (44M12D3), Alexa Fluor™ 594, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP222377AF594
Catalog No. NP222377A94
Only null left
Add to Cart
Description
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Siglec-3/CD33
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
CD33
Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
Primary
Human
IgG2a κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
44M12D3
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Mouse
0.1 mL
945
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.