Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-3/CD33 Antibody (44M12D3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222377FR
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Siglec-3/CD33 | |
Monoclonal | |
DyLight 680 | |
AAH28152 | |
CD33 | |
Amino acids 42-159 of human CD33 were used as the immunogen for the antibody. | |
0.1 mL | |
Immunology | |
945 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
44M12D3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction