Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-3/CD33 Antibody (996813) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11374A
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Siglec-3/CD33 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
996813 | |
Flow Cytometry, CyTOF-ready | |
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-3/CD33, Asp18-His259, Accession # P20138 | |
0.1 mL | |
Immunology | |
945 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction