Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-3/CD33 Antibody (CD33 1C7/1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250290F
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for ELISA.Specifications
Siglec-3/CD33 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CD33 | |
Extracellular portion of Siglec-3/CD33 fused to the Fc portion of human IgG1 | |
0.1 mL | |
Immunology | |
945 | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
CD33 1C7/1 | |
ELISA | |
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction