Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-3/CD33 Antibody (6C5/2), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250292AF594
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.Specifications
Siglec-3/CD33 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CD33 | |
Siglec-3/CD33-Fc recombinant protein | |
0.1 mL | |
945 | |
Store at 4°C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry | |
6C5/2 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction