Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-3/CD33 Antibody (hP67.6 (Gemtuzumab)), PerCP - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275906PCP
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-3/CD33 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
CD33 | |
Gemtuzumab was generated by huminization of the murine P67.6 clone, raised against CD33-transfected FMY9S5 cells of human origin. CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells. | |
0.1 mL | |
Primary | |
Recognises human CD33. | |
Store at 4°C in the dark. |
Flow Cytometry | |
hP67.6 (Gemtuzumab) | |
Flow Cytometry | |
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
Rabbit | |
Protein A purified | |
Immunology | |
945.0 | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction