Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-3/CD33 Human anti-Human, DyLight 350, Clone: hP67.6 (Gemtuzumab), Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275905UV

Catalog No. NBP275905UV


Add to cart

Description

Description

Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

Siglec-3/CD33
hP67.6 (Gemtuzumab)
Flow Cytometry, Immunofluorescence
Conjugated, Purified
CD33
Gemtuzumab was generated by huminization of the murine P67.6 clone, raised against CD33-transfected FMY9S5 cells of human origin. CD33 is a transmembrane receptor expressed on predominantly on cells of the myeloid lineage, but it can also be found on some lymphoid cells.
Protein A purified
Immunology
Primary
945.0
Human
Flow Cytometry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Human
IgG4 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Recognises human CD33.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit