Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-3/CD33 LlamabodyTM VHH His-tag Antibody, R&D Systems™

Llama Monoclonal Antibody
$200.50 - $420.50
Specifications
Antigen | Siglec-3/CD33 |
---|---|
Clone | L008.3.25N |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
LMAB10753SP
![]() |
R&D Systems
LMAB10753SP |
25 μg |
Each for $200.50
|
|
|||||
LMAB1075310
![]() |
R&D Systems
LMAB10753100 |
100 μg |
Each for $420.50
|
|
|||||
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-3/CD33 | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
Mouse myeloma cell line NS0-derived human Siglec-3/CD33, Asp18-His259, Accession # AAA51948 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
L008.3.25N | |
Flow Cytometry | |
Unconjugated | |
Llama | |
Human | |
AAA51948 | |
945 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title