Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-6/CD327 Antibody (767329), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2859H
Description
Siglec-6/CD327 Monoclonal antibody specifically detects Siglec-6/CD327 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Siglec-6/CD327 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 946 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 767329 | |
| Flow Cytometry, CyTOF-ready | |
| CD327 antigen, CD33 antigen-like 1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec-6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327, Gln27-Ser331, Accession # O43699 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction