Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-6/CD327 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32122625UL
Description
Siglec-6/CD327 Polyclonal antibody specifically detects Siglec-6/CD327 in Human samples. It is validated for ImmunofluorescenceSpecifications
Siglec-6/CD327 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
CD327 antigen, CD33 antigen-like 1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec-6 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RVKTRRKKAAQPVQNTDDVNPVMVSGSRGHQHQFQTGIVSDHPAEAGPISEDEQELHYAVLHFHKVQPQEPKVTDTEYSEIKIHK | |
25 μg | |
Immunology | |
946 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction