Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-7/CD328 Antibody (6-434), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237732PECY55
Description
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-7/CD328 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
SIGLEC7 | |
Human dendritic cells. | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody 6-434 recognizes CD328 (Siglec-7), a 75 kDa transmembrane glycoprotein expressed mainly on NK cells, dendritic cells and monocytes. HLDA 8; WS Code ARN-80652 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
6-434 | |
Flow Cytometry | |
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7 | |
Mouse | |
Protein A purified | |
Cardiovascular Biology | |
27036.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction