Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-7/CD328 Antibody, Novus Biologicals™
SDP

Catalog No. NB238728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB238728 100 μg
NB236156 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB238728 Supplier Novus Biologicals Supplier No. NBP321387100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Siglec-7/CD328 Polyclonal antibody specifically detects Siglec-7/CD328 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen Siglec-7/CD328
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ERVEGQKSNRKDYSLTMQSSV
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.