Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-8 Antibody (7C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305751
This item is not returnable.
View return policy
Description
Siglec-8 Monoclonal antibody specifically detects Siglec-8 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunoprecipitationSpecifications
Siglec-8 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1-4 ug/mL, ELISA, Immunoprecipitation | |
CD329 antigen, CDw329, MGC59785, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec-8, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
Extracellular domain of human SIGLEC8 fused with Fc fragment of human IgG1 | |
0.1 mg | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, ELISA, Immunoprecipitation | |
7C9 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
27181 | |
Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction