Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-8 Antibody (837535), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7975APCCY7
Description
Siglec-8 Monoclonal antibody specifically detects Siglec-8 in Human samples. It is validated for Flow CytometrySpecifications
Siglec-8 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27181 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
837535 | |
Flow Cytometry | |
CD329 antigen, CDw329, MGC59785, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec-8, SIGLEC8Lsialic acid-binding Ig-like lectin 8 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-8, Met17-Ala363, Accession # Q9NYZ4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction