Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-9 Antibody (K8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314622
Description
Siglec-9 Monoclonal antibody specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, ImmunoprecipitationSpecifications
Siglec-9 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry 1-4 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
Recombinant Siglec-9-Hu IgGFc fusion protein | |
0.1 mg | |
Cardiovascular Biology | |
27180 | |
Store at 4°C. Do not freeze. | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
K8 | |
Unconjugated | |
PBS, pH 7.4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction