Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-9 Antibody (OTI1D9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274189C
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Siglec-9 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SIGLEC9 | |
Full length recombinant protein of human Siglec-9 (NP_055256) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Canine, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D9 | |
Western Blot, Immunohistochemistry | |
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology | |
27180.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction