Learn More
Description
Specifications
Specifications
| Antigen | Siglec-F |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | E50-2440 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Blue 515 (BB515) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5; CD170 |
| Host Species | Rat |
| Immunogen | Mouse Siglec-F and human IgG Fc recombinant fusion protein |
| Show More |
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.