Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-F Rat anti-Mouse, PerCP-Cy5.5, Clone: E50-2440, BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565526
Description
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow cytometry.
Specifications
Siglec-F | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
E50-2440 | |
PerCP-Cyanine5.5 | |
Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5; CD170 | |
Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with PerCP-Cy5.5 under optimum conditions and unconjugated antibody and free PerCP-Cy5.5 were removed. Storage of PerCP-Cy5.5 conjugates in unoptimized diluent is not recommended and may result in loss of signal intensity. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction