Learn More
Invitrogen™ Siglec-G Monoclonal Antibody (SH2.1), PerCP-eFluor™ 710, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 46583382

Description
Description: The SH2.1 monoclonal antibody reacts with mouse siglec-G, which is also known as sialic acid-binding immunoglobulin-like lectin-G. Siglec-G is an inhibitory receptor that is expressed in a B cell-specific manner with B1 B cells expressing the highest levels of siglec-G. When overexpressed, siglec-G inhibits B cell receptor-mediated calcium signaling. Siglec-G-deficient mice show a massive expansion of the B1a cell population as well as higher titers of natural IgM antibodies. Siglec-G-deficient B1a cells show a lower level of spontaneous apoptosis and a prolonged life span, which may result from higher expression levels of the transcription factor NFATc1. Siglec-G-dependent negative regulation exists in B1 cells, which may explain the naturally muted signaling response of B1 cells. Applications Reported: This SH2.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SH2.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of P...
Siglec-G is an inhibitory receptor that is expressed in a B cell-specific manner with B1 B cells expressing the highest levels of siglec-G. When overexpressed, siglec-G inhibits B cell receptor-mediated calcium signaling. Siglec-G-deficient mice show a massive expansion of the B1a cell population as well as higher titers of natural IgM antibodies. Siglec-G-deficient B1a cells show a lower level of spontaneous apoptosis and a prolonged life span, which may result from higher expression levels of the transcription factor NFATc1. Siglec-G-dependent negative regulation exists in B1 cells, which may explain the naturally muted signaling response of B1 cells.
Specifications
Siglec-G | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q80ZE3 | |
Siglecg | |
Affinity Chromatography | |
RUO | |
243958 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
SH2.1 | |
PerCP-eFluor 710 | |
Siglecg | |
9830164H23; A630096C01Rik; mSiglec-G; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Sialic acid-binding Ig-like lectin G; Siglec; Siglec 10; Siglec10; Siglec-10; Siglecg; Siglec-G | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.