Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-H Antibody (23M14A10), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP227061G
Description
Siglec-H Monoclonal specifically detects Siglec-H in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Siglec-H | |
Monoclonal | |
DyLight 488 | |
Q3Y597 | |
Siglech | |
SkRSHTkRVERPATHMDQA (aa286-304) | |
0.05 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
23M14A10 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
6430529G09Rik, Siglec-H, Siglech sialic acid binding Ig-like lectin H | |
Rat | |
Protein G purified | |
RUO | |
233274 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction