Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-H Mouse, anti-Mouse, R718, Clone: 440C, BD OptiBuild™

Catalog No. BDB751846 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751846 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751846 Supplier BD Biosciences Supplier No. 751846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 440c monoclonal antibody specifically recognizes Siglec-H, a type I transmembrane glycoprotein that is encoded by Siglech (Sialic acid binding Ig-like lectin H). Siglec-H contains two immunoglobulin domains in its extracellular region and a cytoplasmic domain that lacks tyrosine-based signaling motifs unlike other CD33-related Siglec-like molecules. Siglec-H is expressed on progenitors of plasmacytoid dendritic cells (pDCs) and depends on DAP12 for cell surface expression and intracellular signaling function. In addition to its expression on pDC, Siglec-H may also be expressed by splenic marginal zone macrophages, medullary macrophages in lymph nodes, and microglia. Siglec-H may regulate the production of type I interferons (IFN-I) by pDCs and other cell types. There is no clear human ortholog for mouse Siglec-H.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-H
Applications Flow Cytometry
Classification Monoclonal
Clone 440C
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.