Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siglec-H Rat anti-Mouse, Alexa Fluor™ 647, Clone: 551, BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 567806
This item is not returnable.
View return policy
Description
Siglec-H Monoclonal antibody specifically detects Siglec-H in Mouse samples. It is validated for Flow cytometry (Routinely Tested)
Specifications
Siglec-H | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
551 | |
Alexa Fluor 647 | |
Siglec-H; Siglech; sialic acid binding Ig-like lectin H | |
Mouse Extracellular Siglec-H Domain Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction