Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-H, Rat anti-Mouse, Clone: 551, Millipore Sigma™

Catalog No. MABF204525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF204525 25 μg
MABF2045100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF204525 Supplier MilliporeSigma Supplier No. MABF204525UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Siglec-H, clone 551, Cat. No. MABF2045, is a rat monoclonal antibody that detects Sialic acid binding Ig-like lectin H and has been tested for use in Flow Cytometry and Immunohistochemistry.

Siglec-H (UniProt: Q3Y597; also known as Sialic acid binding Ig-like lectin H) is encoded by the Siglech gene (Gene ID: 233274) in murine species. Siglecs are sialic acid-binding immunoglobulin-like lectins that are expressed mostly in the immune system and may be involved in both adhesive and signaling functions. Siglec-H is a type I, CD33-related siglec-like transmembrane glycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off in the mature form. The mature form has to ig-like domains (aa 32-141 and 151-234) in its extracellular region. Unlike other CD33-related siglecs, it lacks tyrosine-based signaling motifs in its cytoplasmic tail. Siglec-H is shown to be expressed on plasmaocytoid dendritic cells (pDCs) and can serve as a reliable marker for these cells. Siglec-H is also reported to serve as an endocytic receptor expressed on murine pDCs. Action of Siglec-H depends on a 12 kDa DNAX-activating protein (DAP). (Ref.: Zhang, J., et al. (2006). Blood 107(9); 3600-3608; Yun, TJ., et al., (2016), Cell Metabolism 23(5); 852-866).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-H
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 551
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A representative lot detected Siglec-H in Immunohistochemistry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).Flow Cytometry Analysis: A representative lot detected Siglec-H in Flow Cytometry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).
Formulation Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Sialic acid binding Ig-like lectin H;Siglech
Gene Symbols Siglech;Siglec-H
Host Species Rat
Immunogen Bone marrow-derived Type 1 interferon-producing cells.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_848821.2
Test Specificity Clone 551 is a rat monoclonal antibody that detects murine Siglec-H. It targets an epitope with in the extracellular domain.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.