Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC10 Monoclonal Antibody (5G6)
GREENER_CHOICE

Catalog No. PIMA528237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528237 Supplier Invitrogen™ Supplier No. MA528237
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Human Siglec-10 antibody, clone 5G6 recognizes human Siglec-10 (Sialic acid-binding Ig-like lectin 10), a putative adhesion molecule and member of the Ig superfamily, expressed by monocytes, B cells, eosinophils, and at a higher level by a subpopulation of CD16+CD56- natural killer (NK) cells.

SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC10
Applications Flow Cytometry
Classification Monoclonal
Clone 5G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene SIGLEC10
Gene Accession No. Q96LC7
Gene Alias PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940
Gene Symbols SIGLEC10
Host Species Mouse
Immunogen Recombinant human Siglec-10, fused with the Fc region of human IgG.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.