Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLEC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139815
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YSEIKIHTGQ PLRGPGFGLQ LEREMSGMVP K.
Siglecs are sialic acid-binding lectins of the immunoglobulin superfamily that are mainly expressed in cells of the hematopoietic system. Siglec11 is unlike other siglecs in that it binds specifically to alpha2-8-linked sialic acids and is not found in peripheral blood leukocytes but instead on macrophages of various tissues. Siglec11 is highly homologous to Siglec10 over their extracellular domain but not over their cytoplasmic domain, suggesting that Siglec11 arose through gene duplication followed by a recombination event involving another ancestral siglec gene. Following treatment of Siglec11-transfected cells with pervanadate, Siglec11 becomes tyrosine-phosphorylated and strongly associates with SHP-1 and SHP-2. At least five isoforms of Siglec11 are known to exist.
Specifications
SIGLEC11 | |
Polyclonal | |
Unconjugated | |
SIGLEC11 | |
sialic acid binding Ig like lectin 11; sialic acid binding Ig-like lectin 11; sialic acid-binding Ig-like lectin 11; Sialic acid-binding lectin 11; Siglec; SIGLEC11; siglec-11; UNQ9222/PRO28718 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96RL6 | |
SIGLEC11 | |
Recombinant protein corresponding to Human SIGLEC11. Recombinant protein control fragment (Product #RP-101402). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction