Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIGLEC12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548763
Description
Sialic acid-binding immunoglobulin-like lectins are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene.
Specifications
SIGLEC12 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96PQ1, Q91Y57 | |
SIGLEC12, Siglece | |
KLH conjugated synthetic peptide between 565-595 amino acids from the C-terminal region of human SIGLEC12. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SIGLEC12 | |
Sialic acid-binding Ig-like lectin 12, Siglec-12, Sialic acid-binding Ig-like lectin-like 1, Siglec-L1, SIGLEC12, SIGLECL1, SLG | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
83382, 89858 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction