Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC14 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5119412

 View more versions of this product

Catalog No. PIPA5119412


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

SIGLEC14 Polyclonal Antibody for Western Blot, ICC/IF

Putative adhesion molecule. Sialic acid-binding paired receptor which may activate associated receptors. Interacts with TYROBP. Mainly expressed in hematopoietic tissues including bone marrow, spleen and fetal liver. Siglec-14 is predominantly expressed in hematopoietic tissues but is also found in lung and testis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIGLEC14
Polyclonal
Unconjugated
SIGLEC14
LLNLR-470E3.1; sialic acid binding Ig like lectin 14; sialic acid binding Ig-like lectin 14; sialic acid-binding Ig-like lectin 14; SIG14; Siglec; SIGLEC14; Siglec-14; UNQ294; UNQ294/PRO333
Rabbit
Antigen affinity chromatography, Protein A
RUO
100049587
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q08ET2
SIGLEC14
Recombinant protein Sialic Acid Binding Ig Like Lectin 14. The antigen corresponds to amino acid range 24-137 of the target protein.
200 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.