Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC14 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985470
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5470 Supplier Bioss Supplier No. BS3525R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Putative adhesion molecule. Sialic acid-binding paired receptor which may activate associated receptors. Interacts with TYROBP. Mainly expressed in hematopoietic tissues including bone marrow, spleen and fetal liver. Siglec-14 is predominantly expressed in hematopoietic tissues but is also found in lung and testis.

Specifications

Antigen SIGLEC14
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIGLEC14
Gene Accession No. Q08ET2
Gene Alias LLNLR-470E3.1; sialic acid binding Ig like lectin 14; sialic acid binding Ig-like lectin 14; Sialic acid-binding Ig-like lectin 14; SIG14; Siglec; SIGLEC14; Siglec-14; UNQ294; UNQ294/PRO333
Gene Symbols SIGLEC14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SIGLEC14.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049587
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.