Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC5 Recombinant Rabbit Monoclonal Antibody (110)
GREENER_CHOICE

Catalog No. PIMA536019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536019 Supplier Invitrogen™ Supplier No. MA536019
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

SIGLEC5 Recombinant Monoclonal Antibody for Flow

SIGLEC5 (CD170) is a member of the sialic acid-binding immunoglobulin-like lectin family expressed mainly on myeloid cells such as neutrophils and monocytes. It functions as an inhibitory receptor that recognizes sialylated glycans and transduces signals through immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in its cytoplasmic tail, helping regulate innate immune responses and dampen inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC5
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 110
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SIGLEC5
Gene Accession No. O15389
Gene Alias CD170
Gene Symbols SIGLEC5
Host Species Rabbit
Immunogen Recombinant Human SIGLEC5 Protein (1-434).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.