Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLEC6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519357
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
Specifications
| SIGLEC6 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC6 | |
| CD327; CD33 antigen-like 1; CD33L; CD33L1; CD33L2; CDW327; OBBP1; OB-BP1; obesity-binding protein 1; sialic acid binding Ig like lectin 6; sialic acid binding Ig-like lectin 6; sialic acid-binding Ig-like lectin 6; Siglec; SIGLEC6; Siglec-6 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 946 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43699 | |
| SIGLEC6 | |
| Synthetic peptide sequence (KWMKYGYTSSK) corresponding to the internal amino acids of SIGLEC6 (aa 126-137). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction